siRNA Details
Gene Symbol: Bloc1s6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019788.3 | NP_062762.1 | 4 | 457 | ||
GenBank mRNA | AK038381.1 | - | 4 | 435 | ||
AK077720.1 | - | 4 | 439 | |||
AK147116.1 | - | 4 | 460 | |||
AK160580.1 | - | 4 | 427 | |||
AK170964.1 | - | 4 | 388 | |||
AK172571.1 | - | 4 | 438 | |||
BC016554.1 | - | 4 | 436 | |||
BC085260.1 | - | 4 | 426 | 6437935 |
