siRNA Details
Gene Symbol: Bloc1s6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019788.3 | NP_062762.1 | 1, 2 | 211 | ||
GenBank mRNA | AK038381.1 | - | 1, 2 | 189 | ||
AK077720.1 | - | 1, 2 | 193 | |||
AK147116.1 | - | 1, 2 | 214 | |||
AK160580.1 | - | 1, 2 | 181 | |||
BY015259.1 | - | 1, 2 | 212 |
