siRNA Details
Gene Symbol: Pak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195046.1 | NP_001181975.1 | 13 | 1327 | ||
NM_001195047.1 | NP_001181976.1 | 12 | 1282 | |||
NM_001195048.1 | NP_001181977.1 | 13 | 1437 | |||
NM_001195049.1 | NP_001181978.1 | 15 | 1701 | |||
NM_008778.3 | NP_032804.2 | 12 | 1310 | |||
GenBank mRNA | AF082297.1 | - | 11 | 1224 | ||
AJ496262.1 | - | 11 | 1192 | |||
AJ496263.1 | - | 12 | 1237 | |||
AK031853.1 | - | 13 | 1318 | |||
AK158893.1 | - | 13 | 1437 | |||
AM943853.1 | - | 12 | 1255 | |||
AM943854.1 | - | 13 | 1300 | |||
BC053403.1 | - | 12 | 1317 | 30060082 | ||
U39738.1 | - | 16 | 1708 |
