siRNA Details
Gene Symbol: Pcnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282992.1 | NP_001269921.1 | 6 | 972 | ||
NM_008787.3 | NP_032813.3 | 6 | 972 | |||
GenBank mRNA | AB207233.1 | - | 6 | 846 | ||
BC085244.1 | - | 6 | 796 | |||
BC100380.1 | - | 6 | 767 | |||
BU518252.1 | - | 493 |
