siRNA Details
Gene Symbol: Pcsk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008792.4 | NP_032818.1 | 3 | 662 | ||
GenBank mRNA | AF483512.1 | - | 3 | 304 | ||
AF483513.1 | - | 3 | 304 | |||
AK133620.1 | - | 3 | 663 | |||
AK158493.1 | - | 3 | 610 | |||
AK158598.1 | - | 3 | 724 | |||
AK161585.1 | - | 3 | 603 | |||
BC052013.1 | - | 3 | 579 | 5707923 | ||
BC057348.1 | - | 3 | 617 | 6827695 | ||
M55669.1 | - | 3 | 388 |
