siRNA Details
Gene Symbol: Pdcd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168491.1 | NP_001161963.1 | 12 | 1581 | ||
NM_001168492.1 | NP_001161964.1 | 12 | 1581 | |||
NM_011050.4 | NP_035180.2 | 12 | 1669 | |||
GenBank mRNA | AK134366.1 | - | 12 | 1670 | ||
AK172654.1 | - | 12 | 1537 | |||
BC055739.1 | - | 12 | 1516 | 6414089 | ||
CT010188.1 | - | 11 | 1379 | |||
D50465.1 | - | 11 | 1449 | |||
D86344.1 | - | 12 | 1465 |
