siRNA Details
Gene Symbol: Pfkm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163487.1 | NP_001156959.1 | 16 | 1590 | ||
NM_001163488.1 | NP_001156960.1 | 16 | 1526 | |||
NM_021514.4 | NP_067489.3 | 15 | 1543 | |||
GenBank mRNA | AF249894.1 | - | 15 | 1483 | ||
AK002711.2 | - | 16 | 1493 | |||
AK049773.1 | - | 16 | 1498 | |||
BC005526.1 | - | 15 | 1507 | 3495173 |
