siRNA Details
Gene Symbol: Prkaca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277898.1 | NP_001264827.1 | 9 | 1003 | ||
NM_008854.5 | NP_032880.1 | 9 | 1046 | |||
GenBank mRNA | AK188175.1 | - | 1 | 27 | ||
BC003238.1 | - | 9 | 1036 | 3497908 | ||
BC054834.1 | - | 9 | 1031 | 6439034 | ||
CD807077.1 | - | 5 | 450 |
