siRNA Details
Gene Symbol: Prkca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011101.3 | NP_035231.2 | 4, 5 | 616 | ||
GenBank mRNA | AK147084.1 | - | 4, 5 | 617 | ||
BC096493.1 | - | 4, 5 | 410 | 30742571 | ||
M25811.1 | - | 4, 5 | 650 | |||
X52684.1 | - | 5, 6 | 557 | |||
X52685.1 | - | 5, 6 | 557 |
