siRNA Details
Gene Symbol: Prkce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011104.3 | NP_035234.1 | 13 | 2654 | ||
GenBank mRNA | AF028009.1 | - | 13 | 1855 | ||
AF325507.1 | - | 13 | 1846 | |||
AK082438.1 | - | 13 | 2655 | |||
AK085351.1 | - | 14 | 2066 |
