siRNA Details
Gene Symbol: Prkci | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008857.3 | NP_032883.2 | 15 | 1660 | ||
GenBank mRNA | AK144979.1 | - | 15 | 1660 | ||
AK147710.1 | - | 15 | 1632 | |||
AK167411.1 | - | 15 | 1629 | |||
AK220517.1 | - | 15 | 1684 | |||
BC021630.1 | - | 15 | 1531 | 3980628 | ||
D28577.1 | - | 15 | 1455 |
