siRNA Details
Gene Symbol: Pkd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013630.2 | NP_038658.2 | 21 | 8288 | ||
GenBank mRNA | AK147429.1 | - | 6 | 967 | ||
AK154998.1 | - | 4 | 777 | |||
U70209.1 | - | 21 | 8290 |
