siRNA Details
Gene Symbol: Pkd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013630.2 | NP_038658.2 | 23 | 8969 | ||
GenBank mRNA | AK147429.1 | - | 8 | 1648 | ||
AK154998.1 | - | 6 | 1458 | |||
AK161088.1 | - | 1 | 1305 | |||
U70209.1 | - | 23 | 8971 |
