siRNA Details
Gene Symbol: Plcb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145830.1 | NP_001139302.1 | 19, 20 | 2401 | ||
NM_019677.2 | NP_062651.2 | 19, 20 | 2401 | |||
GenBank mRNA | AF498249.1 | - | 19, 20 | 2032 | ||
AF498250.1 | - | 19, 20 | 2032 | |||
AK129166.1 | - | 11, 12 | 1266 | |||
AK141164.1 | - | 19, 20 | 2349 | |||
BC058710.1 | - | 19, 20 | 2368 | 6842016 | ||
U85712.1 | - | 19, 20 | 2035 | |||
U85713.1 | - | 19, 20 | 2035 | |||
U85714.1 | - | 19, 20 | 2035 |
