siRNA Details
Gene Symbol: Pld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164056.1 | NP_001157528.1 | 17, 18 | 2141 | ||
NM_008875.4 | NP_032901.2 | 16, 17 | 2086 | |||
GenBank mRNA | AK136111.1 | - | 11, 12 | 1324 | ||
BC068144.1 | - | 16, 17 | 1943 | 30532470 | ||
U87868.1 | - | 17, 18 | 2124 |
