siRNA Details
Gene Symbol: Pou2f3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011139.2 | NP_035269.2 | 7 | 519 | ||
GenBank mRNA | AK153697.1 | - | 609 | |||
L14677.1 | - | 7 | 517 | |||
Z18537.1 | - | 7 | 520 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pou2f3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011139.2 | NP_035269.2 | 7 | 519 | ||
GenBank mRNA | AK153697.1 | - | 609 | |||
L14677.1 | - | 7 | 517 | |||
Z18537.1 | - | 7 | 520 |