siRNA Details
Gene Symbol: Mapk11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011161.5 | NP_035291.4 | 6 | 555 | ||
GenBank mRNA | AF135185.1 | - | 6 | 535 | ||
AK144846.1 | - | 6 | 528 | |||
AK161190.1 | - | 5 | 531 | |||
AK161562.1 | - | 6 | 507 | |||
BC057211.1 | - | 6 | 491 | |||
BC064737.1 | - | 6 | 506 | 30112690 | ||
BC092526.1 | - | 6 | 555 | 6593321 |
