siRNA Details
Gene Symbol: St14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011176.4 | NP_035306.2 | 15 | 1939 | ||
GenBank mRNA | AF042822.3 | - | 15 | 1828 | ||
AK052738.1 | - | 15 | 1939 | |||
AK166745.1 | - | 15 | 1930 | |||
BC005496.1 | - | 15 | 1938 | 3488059 | ||
BC008514.1 | - | 15 | 1938 |
