siRNA Details
Gene Symbol: Psen1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008943.2 | NP_032969.1 | 7 | 1158 | ||
GenBank mRNA | AF149111.1 | - | 5 | 670 | ||
AK134232.1 | - | 7 | 841 | |||
AK134615.1 | - | 7 | 880 | |||
AK148101.1 | - | 7 | 879 | |||
AK165984.1 | - | 7 | 879 | |||
AK169963.1 | - | 3 | 285 | |||
AK178426.1 | - | 1 | 84 | |||
BC014744.1 | - | 7 | 909 | 4511140 | ||
BC030409.1 | - | 7 | 833 | 5133302 | ||
BC071233.1 | - | 7 | 897 | 3673609 | ||
L42177.1 | - | 7 | 854 |
