siRNA Details
Gene Symbol: Prpf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138659.2 | NP_619600.2 | 26 | 4603 | ||
GenBank mRNA | AB047391.1 | - | 26 | 4329 | ||
BC034648.1 | - | 6 | 886 | |||
BC093481.1 | - | 26 | 4533 | 30945329 | ||
CN530184.1 | - | 3 | 503 |
