siRNA Details
Gene Symbol: Prpf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138659.2 | NP_619600.2 | 37 | 6391 | ||
GenBank mRNA | AB047391.1 | - | 37 | 6117 | ||
AK077412.1 | - | 6 | 764 | |||
AK167984.1 | - | 2 | 245 | |||
BC034648.1 | - | 17 | 2674 | |||
BC093481.1 | - | 37 | 6321 | 30945329 | ||
CO426130.1 | - | 5 | 642 | |||
CV561799.1 | - | 1 | 86 |
