siRNA Details
Gene Symbol: Prpf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138659.2 | NP_619600.2 | 38 | 6619 | ||
GenBank mRNA | AB047391.1 | - | 38 | 6345 | ||
AK077412.1 | - | 7 | 992 | |||
AK167984.1 | - | 3 | 473 | |||
BC034648.1 | - | 18 | 2902 | |||
BC093481.1 | - | 38 | 6549 | 30945329 | ||
CV561799.1 | - | 2 | 314 |
