siRNA Details
Gene Symbol: Nlgn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163387.1 | NP_001156859.1 | 6 | 2130 | ||
NM_138666.3 | NP_619607.2 | 8 | 3157 | |||
GenBank mRNA | AK122433.1 | - | 8 | 3158 | ||
AK138209.1 | - | 3 | 991 | |||
BC098461.1 | - | 6 | 2130 | 30099449 |
