siRNA Details
Gene Symbol: Flna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290421.1 | NP_001277350.1 | 35 | 5803 | ||
NM_010227.3 | NP_034357.2 | 34 | 5781 | |||
GenBank mRNA | AK131179.1 | - | 22 | 3622 | ||
BC038478.1 | - | 35 | 5678 | |||
BC039208.1 | - | 14 | 1993 | |||
BC151024.1 | - | 34 | 5772 | 9087937 |
