siRNA Details
Gene Symbol: Ptger4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136079.2 | NP_001129551.1 | 2 | 995 | ||
NM_008965.2 | NP_032991.1 | 2 | 932 | |||
GenBank mRNA | AK145045.1 | - | 2 | 932 | ||
AK156428.1 | - | 2 | 934 | |||
BC009023.1 | - | 2 | 490 | 3481696 | ||
BC011193.1 | - | 2 | 980 | 4037704 | ||
D13458.1 | - | 2 | 576 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
