siRNA Details
Gene Symbol: Tmlhe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138758.1 | NP_620097.1 | 7, 8 | 1259 | ||
GenBank mRNA | AK144627.1 | - | 7, 8 | 1255 | ||
AK161724.1 | - | 7, 8 | 1226 | |||
AY033513.1 | - | 6, 7 | 1097 | |||
BC010495.1 | - | 7, 8 | 1259 | 4195680 | ||
BC115365.1 | - | 7, 8 | 1191 | 40059106 |
