siRNA Details
Gene Symbol: Ptpn13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011204.2 | NP_035334.2 | 22 | 3724 | ||
GenBank mRNA | AK014577.1 | - | 21 | 3193 | ||
D28529.1 | - | 2 | 626 | |||
D83966.1 | - | 22 | 3412 | |||
Z32740.1 | - | 21 | 3381 |
