siRNA Details
Gene Symbol: Ptpn13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011204.2 | NP_035334.2 | 37, 38 | 6290 | ||
GenBank mRNA | AK161200.1 | - | 7, 8 | 923 | ||
D83966.1 | - | 37, 38 | 5978 | |||
Z32740.1 | - | 36, 37 | 5944 |
