siRNA Details
Gene Symbol: Ptprb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029928.2 | NP_084204.2 | 8 | 1962 | ||
GenBank mRNA | AK147439.1 | - | 8 | 1964 | ||
AK147668.1 | - | 8 | 1964 | |||
AY077755.1 | - | 8 | 1840 | |||
BC141006.1 | - | 8 | 1758 | 9055618 | ||
BC145111.1 | - | 8 | 1758 | 9053659 |
