siRNA Details
Gene Symbol: Rab23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159729.1 | NP_001153201.1 | 3, 4 | 489 | ||
NM_008999.4 | NP_033025.2 | 3, 4 | 601 | |||
GenBank mRNA | AB232618.1 | - | 1, 2 | 166 | ||
AK016493.1 | - | 3, 4 | 501 | |||
AK045454.1 | - | 3, 4 | 360 | |||
AK047068.1 | - | 3, 4 | 602 | |||
AK054079.1 | - | 3 | 1515 | |||
AK082017.1 | - | 3, 4 | 564 | |||
AK163909.1 | - | 3, 4 | 610 | |||
AK164057.1 | - | 3, 4 | 685 | |||
M79305.1 | - | 1, 2 | 106 | |||
Z22821.1 | - | 1, 2 | 186 |
