siRNA Details
Gene Symbol: Rad51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011234.4 | NP_035364.1 | 7 | 778 | ||
GenBank mRNA | AK011242.1 | - | 7 | 779 | ||
AK045050.1 | - | 7 | 781 | |||
AK076468.1 | - | 7 | 726 | |||
AK148165.1 | - | 778 | ||||
AK151157.1 | - | 7 | 757 | |||
AK151177.1 | - | 7 | 757 | |||
AK160282.1 | - | 7 | 770 | |||
BC027384.1 | - | 7 | 743 | 4951015 | ||
D13473.1 | - | 7 | 733 | |||
D13803.1 | - | 7 | 746 |
