siRNA Details
Gene Symbol: Rad51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011234.4 | NP_035364.1 | 5 | 617 | ||
GenBank mRNA | AK011242.1 | - | 5 | 618 | ||
AK045050.1 | - | 5 | 620 | |||
AK076468.1 | - | 5 | 565 | |||
AK148165.1 | - | 617 | ||||
AK151157.1 | - | 5 | 596 | |||
AK151177.1 | - | 5 | 596 | |||
AK160282.1 | - | 5 | 609 | |||
BC027384.1 | - | 5 | 582 | 4951015 | ||
D13473.1 | - | 5 | 572 | |||
D13803.1 | - | 5 | 585 |
