siRNA Details
Gene Symbol: Rad51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011234.4 | NP_035364.1 | 7, 8 | 863 | ||
GenBank mRNA | AK011242.1 | - | 7, 8 | 864 | ||
AK076468.1 | - | 7, 8 | 811 | |||
AK151157.1 | - | 7, 8 | 842 | |||
AK151177.1 | - | 7, 8 | 842 | |||
AK160282.1 | - | 7, 8 | 855 | |||
BC027384.1 | - | 7, 8 | 828 | 4951015 | ||
D13473.1 | - | 7, 8 | 818 | |||
D13803.1 | - | 7, 8 | 831 |
