siRNA Details
Gene Symbol: Rbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282000.1 | NP_001268929.1 | 16 | 2594 | ||
NM_001282001.1 | NP_001268930.1 | 17 | 2693 | |||
NM_011250.4 | NP_035380.3 | 17 | 2723 | |||
GenBank mRNA | AK160027.1 | - | 17 | 2670 | ||
BC020528.1 | - | 17 | 2671 | 4240592 | ||
U36799.1 | - | 17 | 2593 | |||
U47333.1 | - | 16 | 2536 | |||
U50850.1 | - | 17 | 2631 |
