siRNA Details
Gene Symbol: Reln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011261.2 | NP_035391.2 | 57 | 9639 | ||
GenBank mRNA | AK017094.1 | - | 2 | 212 | ||
AK047608.1 | - | 11 | 1870 | |||
BC118041.1 | - | 11 | 1788 | |||
D63520.1 | - | 16 | 2889 | |||
U24703.1 | - | 57 | 9624 |
