siRNA Details
Gene Symbol: Rfx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166414.1 | NP_001159886.1 | 5 | 569 | ||
NM_011265.3 | NP_035395.2 | 4 | 610 | |||
GenBank mRNA | AK030008.1 | - | 6 | 642 | ||
AK041120.1 | - | 5 | 670 | |||
AK140928.1 | - | 4 | 493 | |||
BC017598.1 | - | 4 | 584 | 4483833 |
