siRNA Details
Gene Symbol: Trim10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011280.2 | NP_035410.2 | 2, 3 | 824 | ||
GenBank mRNA | AF134811.1 | - | 2, 3 | 806 | ||
AF220121.1 | - | 2, 3 | 811 | |||
AK011082.1 | - | 2, 3 | 824 | |||
BC051632.1 | - | 2, 3 | 797 | 6507331 |
