siRNA Details
Gene Symbol: Rock2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009072.2 | NP_033098.2 | 16 | 2121 | ||
GenBank mRNA | AK035509.1 | - | 1 | 14 | ||
AK144687.1 | - | 16 | 1975 | |||
AK172983.1 | - | 1 | 76 | |||
DQ864977.1 | - | 16 | 1932 | |||
U58513.1 | - | 16 | 1909 |
