siRNA Details
Gene Symbol: Rock2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009072.2 | NP_033098.2 | 5 | 750 | ||
GenBank mRNA | AK144687.1 | - | 5 | 604 | ||
AK150181.1 | - | 5 | 604 | |||
AK163082.1 | - | 5 | 604 | |||
BC029113.1 | - | 3 | 331 | |||
DQ864977.1 | - | 5 | 561 | |||
U58513.1 | - | 5 | 538 |
