siRNA Details
Gene Symbol: Rock2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009072.2 | NP_033098.2 | 20 | 2613 | ||
GenBank mRNA | AK035509.1 | - | 5 | 506 | ||
AK045517.1 | - | 4 | 375 | |||
AK144687.1 | - | 20 | 2467 | |||
AK172983.1 | - | 5 | 568 | |||
DQ864977.1 | - | 20 | 2424 | |||
U58513.1 | - | 20 | 2401 |
