siRNA Details
Gene Symbol: Slc22a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164634.1 | NP_001158106.1 | 2 | 517 | ||
NM_001164635.1 | NP_001158107.1 | 2 | 425 | |||
NM_031194.5 | NP_112471.3 | 3 | 597 | |||
GenBank mRNA | AB079895.1 | - | 1 | 254 | ||
AF078869.1 | - | 2 | 372 | |||
AK031962.1 | - | 3 | 563 | |||
AK044336.1 | - | 2 | 394 | |||
AK157891.1 | - | 2 | 426 | |||
BB622154.1 | - | 3 | 560 | |||
BC014762.1 | - | 2 | 403 | 4239544 | ||
BU615016.1 | - | 2 | 467 |
