siRNA Details
Gene Symbol: Rp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011283.2 | NP_035413.1 | 4 | 4809 | ||
GenBank mRNA | AF146593.2 | - | 1 | 1475 | ||
AF155141.2 | - | 4 | 4800 | |||
BC028267.1 | - | 1 | 1359 | |||
BC038794.1 | - | 4 | 4750 | |||
BC061512.1 | - | 1 | 2030 | |||
BC096607.1 | - | 4 | 4809 | |||
BC120927.1 | - | 4 | 4763 | 40099705 | ||
BC120928.1 | - | 4 | 4763 | 40099706 |
