siRNA Details
Gene Symbol: Rpgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177950.1 | NP_001171421.1 | 11 | 1566 | ||
NM_001177951.1 | NP_001171422.1 | 11 | 1566 | |||
NM_001177952.1 | NP_001171423.1 | 11 | 1560 | |||
NM_001177953.1 | NP_001171424.1 | 11 | 1566 | |||
NM_011285.2 | NP_035415.1 | 11 | 1566 | |||
GenBank mRNA | AF044677.1 | - | 11 | 1484 | ||
AJ238396.1 | - | 11 | 1478 | |||
AK017192.1 | - | 11 | 1498 | |||
AK029864.1 | - | 11 | 1692 | |||
AK038652.1 | - | 11 | 1530 | |||
AK136298.1 | - | 11 | 1497 | |||
AK138977.1 | - | 11 | 1561 | |||
BC012661.1 | - | 11 | 1505 |
