siRNA Details
Gene Symbol: Ryr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023868.2 | NP_076357.2 | 15 | 1951 | ||
GenBank mRNA | AF295105.1 | - | 15 | 1452 | ||
BC043140.1 | - | 14 | 1930 | |||
BC059061.1 | - | 14 | 1930 |
