siRNA Details
Gene Symbol: Sars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204979.1 | NP_001191908.1 | 5 | 615 | ||
NM_011319.3 | NP_035449.1 | 5 | 615 | |||
GenBank mRNA | AK075827.1 | - | 5 | 546 | ||
AK077309.1 | - | 5 | 497 | |||
AK082793.1 | - | 5 | 526 | |||
AK152150.1 | - | 5 | 573 | |||
AK153158.1 | - | 5 | 565 | |||
AK181805.1 | - | 1 | 20 | |||
AK190309.1 | - | 1 | 20 | |||
AK217331.1 | - | 1 | 20 | |||
AK219731.1 | - | 1 | 20 | |||
BC008612.1 | - | 5 | 504 | 3257101 | ||
BC031932.1 | - | 5 | 504 | |||
M74012.1 | - | 5 | 567 |
