siRNA Details
Gene Symbol: Scn10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205321.1 | NP_001192250.1 | 13 | 1896 | ||
NM_009134.3 | NP_033160.2 | 13 | 1896 | |||
GenBank mRNA | AK135787.1 | - | 10 | 1517 | ||
AK157866.1 | - | 13 | 1896 | |||
AY538273.1 | - | 12 | 1826 | |||
Y09108.1 | - | 12 | 1861 |
