siRNA Details
Gene Symbol: Sema5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009154.2 | NP_033180.2 | 8 | 1272 | ||
GenBank mRNA | AK031231.1 | - | 8 | 1265 | ||
AK043386.1 | - | 8 | 984 | |||
AK045236.1 | - | 8 | 1276 | |||
AK046502.1 | - | 8 | 1048 | |||
AK141612.1 | - | 8 | 1216 | |||
AK143026.1 | - | 8 | 1283 | |||
AK157706.1 | - | 8 | 1255 | |||
AK165206.1 | - | 7 | 3008 | |||
BC065137.1 | - | 7 | 741 | 30548165 | ||
X97817.1 | - | 6 | 641 |
