siRNA Details
Gene Symbol: Sema6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018744.2 | NP_061214.2 | 6 | 1054 | ||
GenBank mRNA | AF030430.1 | - | 6 | 458 | ||
AF288666.1 | - | 5 | 385 | |||
AK042751.1 | - | 6 | 1055 | |||
AK082711.1 | - | 6 | 1057 | |||
BC059238.1 | - | 6 | 1016 | 6417475 | ||
BC062979.1 | - | 6 | 632 | 6841689 |
