siRNA Details
Gene Symbol: Zfp106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011743.2 | NP_035873.2 | 6 | 2937 | ||
GenBank mRNA | AK147887.1 | - | 6 | 2938 | ||
BC010440.1 | - | 4 | 2579 | |||
BC025424.1 | - | 6 | 2926 | 4459713 | ||
BU695141.1 | - | 2 | 304 |
