siRNA Details
Gene Symbol: Shcbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011369.2 | NP_035499.1 | 1 | 39 | ||
GenBank mRNA | AF017152.1 | - | 1 | 39 | ||
AK051205.1 | - | 1 | 73 | |||
AK052944.1 | - | 1 | 65 | |||
AK137496.1 | - | 1 | 55 | |||
AK144966.1 | - | 1 | 62 | |||
AK149580.1 | - | 1 | 56 | |||
AK162943.1 | - | 1 | 53 | |||
BC070455.1 | - | 1 | 59 | 30606785 |
